C12H316CF4E7 Chemcia Scientific, LLC

3-(1-Benzyloxycarbonylamino-ethyl)-azetidine-1-carboxylic acid tert-butyl ester

  • Molecular Weight : 334.41892

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=10169

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10169
Availability details for
Quantity
  • Availability
  • Add to Cart