C12H316CF4ED Chemcia Scientific, LLC

(4-Trifluoromethyl-pyrrolidin-3-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD24545366
  • Cas Number : 168545-06-0
  • Molecular Weight : 254.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (4-(trifluoromethyl)pyrrolidin-3-yl)carbamate
Inchi Code 1S/C10H17F3N2O2/c1-9(2,3)17-8(16)15-7-5-14-4-6(7)10(11,12)13/h6-7,14H,4-5H2,1-3H3,(H,15,16)
Inchi Key TTZLWOFPFKRYBC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10276

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10276
Availability details for
Quantity
  • Availability
  • Add to Cart