C12H316CF4EF Chemcia Scientific, LLC

3-[(2-Methyl-propane-2-sulfinylimino)-methyl]-azetidine-1-carboxylic acid tert-butyl ester

  • Cas Number : 1426421-85-3
  • Molecular Weight : 288.41183

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10281

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10281
Availability details for
Quantity
  • Availability
  • Add to Cart