C12H316CF4F3 Chemcia Scientific, LLC

3-Amino-6-chloro-2-iodo-benzoic acid methyl ester

  • Cas Number : 2090514-10-4
  • Molecular Weight : 311.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-amino-6-chloro-2-iodobenzoate
Inchi Code 1S/C8H7ClINO2/c1-13-8(12)6-4(9)2-3-5(11)7(6)10/h2-3H,11H2,1H3
Inchi Key HNBUNUGKQKZCNY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10284

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10284
Availability details for
Quantity
  • Availability
  • Add to Cart