C12H316CF4F4 Chemcia Scientific, LLC

(3-Chloro-2-methyl-phenyl)-dimethyl-amine

  • Cas Number : 67761-88-0
  • Molecular Weight : 169.65569

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10285

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10285
Availability details for
Quantity
  • Availability
  • Add to Cart