C12H316CF4F5 Chemcia Scientific, LLC

3-Carbamoyl-acrylic acid methyl ester

  • Molecular Weight : 129.11644

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10286

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10286
Availability details for
Quantity
  • Availability
  • Add to Cart