C12H316CF4F8 Chemcia Scientific, LLC

Methyl-(2-methyl-3-nitro-phenyl)-amine

  • MDL number MFCD17284869
  • Cas Number : 848480-17-1
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N,2-dimethyl-3-nitroaniline
Inchi Code 1S/C8H10N2O2/c1-6-7(9-2)4-3-5-8(6)10(11)12/h3-5,9H,1-2H3
Inchi Key ACASHMOCWAHWMO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10327

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10327
Availability details for
Quantity
  • Availability
  • Add to Cart