C12H316CF4F9 Chemcia Scientific, LLC

2,N-Dimethyl-benzene-1,3-diamine

  • MDL number MFCD17284916
  • Cas Number : 848480-16-0
  • Molecular Weight : 136.19824

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10328

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10328
Availability details for
Quantity
  • Availability
  • Add to Cart