C12H316CF4FF Chemcia Scientific, LLC

6-Benzyloxy-3-methyl-1H-indazole

  • MDL number MFCD30295216
  • Cas Number : 362512-26-3
  • Molecular Weight : 238.29163

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10374

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10374
Availability details for
Quantity
  • Availability
  • Add to Cart