C12H316CF501 Chemcia Scientific, LLC

6-Benzyloxy-3-methyl-indazole-1-carboxylic acid tert-butyl ester

  • Cas Number : 753922-08-6
  • Molecular Weight : 338.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 6-(benzyloxy)-3-methyl-1H-indazole-1-carboxylate
Inchi Code 1S/C20H22N2O3/c1-14-17-11-10-16(24-13-15-8-6-5-7-9-15)12-18(17)22(21-14)19(23)25-20(2,3)4/h5-12H,13H2,1-4H3
Inchi Key OHUGEITWYFXHTH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10376

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10376
Availability details for
Quantity
  • Availability
  • Add to Cart