C12H316CF502 Chemcia Scientific, LLC

2-Fluoro-4-hydrazino-5-methyl-benzonitrile

  • Molecular Weight : 165.17146

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10377

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10377
Availability details for
Quantity
  • Availability
  • Add to Cart