C12H316CF503 Chemcia Scientific, LLC

Dimethyl-(2-methyl-3-nitro-phenyl)-amine

  • MDL number MFCD28363962
  • Cas Number : 183555-60-4
  • Molecular Weight : 180.20819

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Orange Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10378

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10378
Availability details for
Quantity
  • Availability
  • Add to Cart