C12H316CF506 Chemcia Scientific, LLC

4-Methoxy-2-trifluoromethyl-1H-benzoimidazole

  • MDL number MFCD24395801
  • Cas Number : 392-38-1
  • Molecular Weight : 216.16414

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10424

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10424
Availability details for
Quantity
  • Availability
  • Add to Cart