C12H316CF50B Chemcia Scientific, LLC

6-Amino-2-bromo-pyridin-3-ol

  • MDL number MFCD12913806
  • Cas Number : 1187930-22-8
  • Molecular Weight : 189.0124

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10513

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10513
Availability details for
Quantity
  • Availability
  • Add to Cart