C12H316CF50C Chemcia Scientific, LLC

6-Fluoro-5-methyl-2H-indazol-3-ylamine

  • Molecular Weight : 165.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluoro-5-methyl-2H-indazol-3-amine
Inchi Code 1S/C8H8FN3/c1-4-2-5-7(3-6(4)9)11-12-8(5)10/h2-3H,1H3,(H3,10,11,12)
Inchi Key XLAFKBXUNAYWQC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10521

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10521
Availability details for
Quantity
  • Availability
  • Add to Cart