C12H316CF512 Chemcia Scientific, LLC

3,5-Diiodo-6-trifluoromethyl-pyridin-2-ylamine

  • Cas Number : 1824090-38-1
  • Molecular Weight : 413.90821

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10526

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10526
Availability details for
Quantity
  • Availability
  • Add to Cart