C12H316CF513 Chemcia Scientific, LLC

Bis-(4-iodo-phenyl)-diazene

  • Cas Number : 1601-97-4
  • Molecular Weight : 434.01976

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Golden Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10527

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10527
Availability details for
Quantity
  • Availability
  • Add to Cart