C12H316CF51A Chemcia Scientific, LLC

4-Amino-3-chloro-5-nitro-benzoic acid methyl ester

  • MDL number MFCD09842615
  • Cas Number : 863886-04-8
  • Molecular Weight : 230.60899

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10571

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10571
Availability details for
Quantity
  • Availability
  • Add to Cart