C12H316CF51C Chemcia Scientific, LLC

1-(3-Bromo-5-hydroxy-phenyl)-ethanone

  • MDL number MFCD22544402
  • Cas Number : 867366-86-7
  • Molecular Weight : 215.04779

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10586

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10586
Availability details for
Quantity
  • Availability
  • Add to Cart