C12H316CF51D Chemcia Scientific, LLC

1-(5-Chloro-pyrazolo[3,4-c]pyridin-1-yl)-ethanone

  • MDL number MFCD08061149
  • Cas Number : 76006-04-7
  • Molecular Weight : 195.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-acetyl-5-chloro-1H-pyrazolo[3,4-c]pyridine
Inchi Code 1S/C8H6ClN3O/c1-5(13)12-7-4-10-8(9)2-6(7)3-11-12/h2-4H,1H3
Inchi Key MBQYZUMHHMYCBJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Black Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10625

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10625
Availability details for
Quantity
  • Availability
  • Add to Cart