C12H316CF51E Chemcia Scientific, LLC

3-Chloro-2-(4-methoxy-benzyloxy)-benzonitrile

  • Molecular Weight : 273.72139

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10637

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10637
Availability details for
Quantity
  • Availability
  • Add to Cart