C12H316CF520 Chemcia Scientific, LLC

(2-Oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-3-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD19441845
  • Cas Number : 86499-69-6
  • Molecular Weight : 276.33825

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=82

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=82
Availability details for
Quantity
  • Availability
  • Add to Cart