C12H316CF524 Chemcia Scientific, LLC

4-Bromo-2-methoxy-5-nitro-phenylamine

  • MDL number MFCD23703340
  • Cas Number : 1421371-97-2
  • Molecular Weight : 247.04944

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown-Black Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10768

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10768
Availability details for
Quantity
  • Availability
  • Add to Cart