C12H316CF527 Chemcia Scientific, LLC

6-Amino-2,4-dibromo-3-methyl-benzoic acid

  • Molecular Weight : 308.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-2,4-dibromo-3-methylbenzoic acid
Inchi Code 1S/C8H7Br2NO2/c1-3-4(9)2-5(11)6(7(3)10)8(12)13/h2H,11H2,1H3,(H,12,13)
Inchi Key BBFQKVWSPXYXFV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10771

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10771
Availability details for
Quantity
  • Availability
  • Add to Cart