C12H316CF52C Chemcia Scientific, LLC

3-Amino-4-fluoro-2-methyl-benzoic acid methyl ester

  • MDL number MFCD24643109
  • Cas Number : 1079993-18-2
  • Molecular Weight : 183.18395

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Orange Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10773

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10773
Availability details for
Quantity
  • Availability
  • Add to Cart