C12H316CF52D Chemcia Scientific, LLC

1-Chloro-4,5-difluoro-2-methoxy-benzene

  • MDL number MFCD18415542
  • Cas Number : 1261598-22-4
  • Molecular Weight : 178.5671

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10774

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10774
Availability details for
Quantity
  • Availability
  • Add to Cart