C12H316CF531 Chemcia Scientific, LLC

3-Bromo-4-chloro-benzo[b]thiophene-2-carboxylic acid methyl ester

  • Cas Number : 160383-18-6
  • Molecular Weight : 305.57912

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10783

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10783
Availability details for
Quantity
  • Availability
  • Add to Cart