C12H316CF537 Chemcia Scientific, LLC

4-Chloro-5-methyl-thieno[2,3-d]pyrimidine-6-carboxylic acid methyl ester

  • MDL number MFCD05864295
  • Cas Number : 832113-96-9
  • Molecular Weight : 242.68534

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10789

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10789
Availability details for
Quantity
  • Availability
  • Add to Cart