C12H316CF538 Chemcia Scientific, LLC

(5-Fluoro-pyridin-3-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD18968282
  • Cas Number : 342603-20-7
  • Molecular Weight : 212.22571

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10790

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10790
Availability details for
Quantity
  • Availability
  • Add to Cart