C12H316CF53B Chemcia Scientific, LLC

2-Methyl-imidazo[1,2-a]pyridine-6-carboxylic acid

  • MDL number MFCD09258863
  • Cas Number : 129912-22-7
  • Molecular Weight : 176.17631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10793

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10793
Availability details for
Quantity
  • Availability
  • Add to Cart