C12H316CF540 Chemcia Scientific, LLC

2-Formyl-5-methoxy-benzoic acid

  • MDL number MFCD06739603
  • Cas Number : 4785-56-2
  • Molecular Weight : 180.16171

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10860

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10860
Availability details for
Quantity
  • Availability
  • Add to Cart