C12H316CF543 Chemcia Scientific, LLC

3-Iodo-5-nitro-2H-indazole

  • Molecular Weight : 289.03323

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10867

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10867
Availability details for
Quantity
  • Availability
  • Add to Cart