C12H316CF548 Chemcia Scientific, LLC

1-Bromo-5-iodo-4-methoxy-2-trifluoromethyl-benzene

  • Molecular Weight : 380.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-5-iodo-4-methoxy-2-(trifluoromethyl)benzene
Inchi Code 1S/C8H5BrF3IO/c1-14-7-2-4(8(10,11)12)5(9)3-6(7)13/h2-3H,1H3
Inchi Key ZTZTUJMMOUHHEH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10872

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10872
Availability details for
Quantity
  • Availability
  • Add to Cart