C12H316CF54A Chemcia Scientific, LLC

2-Dibromomethyl-6-nitro-benzoic acid methyl ester

  • Molecular Weight : 352.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-(dibromomethyl)-6-nitrobenzoate
Inchi Code 1S/C9H7Br2NO4/c1-16-9(13)7-5(8(10)11)3-2-4-6(7)12(14)15/h2-4,8H,1H3
Inchi Key NPEIYSBHBFHZAD-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10874

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10874
Availability details for
Quantity
  • Availability
  • Add to Cart