C12H316CF54C Chemcia Scientific, LLC

2-Chloro-4-nitro-benzoic acid tert-butyl ester

  • Molecular Weight : 257.67559

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10876

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10876
Availability details for
Quantity
  • Availability
  • Add to Cart