C12H316CF54F Chemcia Scientific, LLC

2-Chloromethyl-1-(4-methoxy-benzyl)-piperidine

  • Cas Number : 1275944-66-5
  • Molecular Weight : 253.7746

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10879

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10879
Availability details for
Quantity
  • Availability
  • Add to Cart