C12H316CF553 Chemcia Scientific, LLC

6-Amino-5-nitro-pyridine-2-carbonitrile

  • MDL number MFCD18072571
  • Cas Number : 516481-67-7
  • Molecular Weight : 164.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-5-nitro-2-pyridinecarbonitrile
Inchi Code 1S/C6H4N4O2/c7-3-4-1-2-5(10(11)12)6(8)9-4/h1-2H,(H2,8,9)
Inchi Key RRPKOJDIMOBIOE-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10907

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10907
Availability details for
Quantity
  • Availability
  • Add to Cart