C12H316CF554 Chemcia Scientific, LLC

5-Chloro-2-methoxy-3-nitro-pyridine

  • MDL number MFCD07375032
  • Cas Number : 22353-52-2
  • Molecular Weight : 188.57135

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10911

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10911
Availability details for
Quantity
  • Availability
  • Add to Cart