C12H316CF555 Chemcia Scientific, LLC

5-Chloro-2-methoxy-pyridin-3-ylamine

  • MDL number MFCD07375045
  • Cas Number : 886373-70-2
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-methoxy-3-pyridinamine
Inchi Code 1S/C6H7ClN2O/c1-10-6-5(8)2-4(7)3-9-6/h2-3H,8H2,1H3
Inchi Key YOVZLBYUUJGCEB-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10914

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10914
Availability details for
Quantity
  • Availability
  • Add to Cart