C12H316CF556 Chemcia Scientific, LLC

6-Amino-5-methoxy-nicotinonitrile

  • MDL number MFCD18261105
  • Cas Number : 1256821-98-3
  • Molecular Weight : 149.15334

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10921

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10921
Availability details for
Quantity
  • Availability
  • Add to Cart