C12H316CF568 Chemcia Scientific, LLC

1-Bromo-2-methoxy-4-thiocyanato-benzene

  • Molecular Weight : 244.11112

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10963

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10963
Availability details for
Quantity
  • Availability
  • Add to Cart