C12H316CF569 Chemcia Scientific, LLC

1-Chloro-2-methoxy-4-thiocyanato-benzene

  • Molecular Weight : 199.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-chloro-2-methoxy-4-thiocyanatobenzene
Inchi Code 1S/C8H6ClNOS/c1-11-8-4-6(12-5-10)2-3-7(8)9/h2-4H,1H3
Inchi Key IKKMCMMQNCJMSA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10964

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10964
Availability details for
Quantity
  • Availability
  • Add to Cart