C12H316CF56E Chemcia Scientific, LLC

2-Chloro-4-fluoro-5-trifluoromethyl-phenylamine

  • MDL number MFCD28556866
  • Cas Number : 125508-25-0
  • Molecular Weight : 213.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-4-fluoro-5-(trifluoromethyl)aniline
Inchi Code 1S/C7H4ClF4N/c8-4-2-5(9)3(1-6(4)13)7(10,11)12/h1-2H,13H2
Inchi Key WAHMPPLIGCOKGX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10999

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10999
Availability details for
Quantity
  • Availability
  • Add to Cart