C12H316CF56F Chemcia Scientific, LLC

(4-Acetyl-pyridin-2-yl)-carbamic acid tert-butyl ester

  • Molecular Weight : 236.27292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11000

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11000
Availability details for
Quantity
  • Availability
  • Add to Cart