C12H316CF574 Chemcia Scientific, LLC

4-Fluoro-2-iodo-5-trifluoromethyl-phenylamine

  • MDL number MFCD11111781
  • Cas Number : 868692-49-3
  • Molecular Weight : 305.01463

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11005

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11005
Availability details for
Quantity
  • Availability
  • Add to Cart