C12H316CF575 Chemcia Scientific, LLC

5-Dimethylamino-2-methyl-benzoic acid methyl ester

  • Molecular Weight : 193.2477

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Red Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11006

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11006
Availability details for
Quantity
  • Availability
  • Add to Cart