C12H316CF576 Chemcia Scientific, LLC

2-Methyl-5-methylamino-benzoic acid methyl ester

  • Cas Number : 1850319-28-6
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-methyl-5-(methylamino)benzoate
Inchi Code 1S/C10H13NO2/c1-7-4-5-8(11-2)6-9(7)10(12)13-3/h4-6,11H,1-3H3
Inchi Key CSHZLNULUOWLOA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Red Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11007

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11007
Availability details for
Quantity
  • Availability
  • Add to Cart