C12H316CF578 Chemcia Scientific, LLC

4-Bromo-quinoline-8-carbonitrile

  • MDL number MFCD08063212
  • Cas Number : 1020743-28-5
  • Molecular Weight : 233.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-8-quinolinecarbonitrile
Inchi Code 1S/C10H5BrN2/c11-9-4-5-13-10-7(6-12)2-1-3-8(9)10/h1-5H
Inchi Key MQKWVLSWSZHDGR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11021

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11021
Availability details for
Quantity
  • Availability
  • Add to Cart