C12H316CF57E Chemcia Scientific, LLC

5-Bromo-4-methyl-2-nitro-phenol

  • MDL number MFCD19374210
  • Cas Number : 861315-14-2
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-4-methyl-2-nitrophenol
Inchi Code 1S/C7H6BrNO3/c1-4-2-6(9(11)12)7(10)3-5(4)8/h2-3,10H,1H3
Inchi Key GFMIIMRPQYYMLY-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11174

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11174
Availability details for
Quantity
  • Availability
  • Add to Cart