C12H316CF580 Chemcia Scientific, LLC

5-Fluoro-2-hydrazino-benzoic acid hydrochloride

  • Molecular Weight : 206.60541

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Salt Data HCl
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11194

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11194
Availability details for
Quantity
  • Availability
  • Add to Cart